missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | N-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride |
| CAS | 2512203-91-5 |
| Analyte or Component Names | N-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21H28ClN3S • 2HCl |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-N,N',N'-trimethyl-1,3-propanediamine Dihydrochloride, N1-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-N1,N3,N3-trimethyl-1,3-propanediamine Dihydrochloride, Chlorpromazine Dihydrochloride Impurity B, Chlorpromazine Dihydrochloride EP Impurity B |
| InChI Formula | InChI=1S/C21H28ClN3S.2ClH/c1-23(2)12-6-13-24(3)14-7-15-25-18-8-4-5-9-20(18)26-21-11-10-17(22)16-19(21)25;;/h4-5,8-11,16H,6-7,12-15H2,1-3H3;2*1H |
| SMILES | ClC1=CC2=C(C=C1)SC3=CC=CC=C3N2CCCN(C)CCCN(C)C |
| IUPAC Name | N1-(3-(2-chloro-10H-phenothiazin-10-yl)propyl)-N1,N3,N3-trimethylpropane-1,3-diamine hydrogen chloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction