missing translation for 'onlineSavingsMsg'
Learn More

HspA6 Antibody [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30513425 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30513425 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30513425 Supplier Novus Biologicals Supplier No. NBP338454UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HspA6 Polyclonal antibody specifically detects HspA6 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HspA6
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B'
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 474-643 of human HspA6 (NP_002146.2).,, Sequence:, PQIEVTFDIDANGILSVTATDRSTGKANKITITNDKGRLSKEEVERMVHEAEQYKAEDEAQRDRVAAKNSLEAHVFHVKGSLQEESLRDKIPEEDRRKMQDKCREVLAWLEHNQLAEKEEYEHQKRELEQICRPIFSRLYGGPGVPGGSSCGTQARQGDPSTGPIIEEVD
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3310
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.