missing translation for 'onlineSavingsMsg'
Learn More

HTF9C Antibody [DyLight 550], Novus Biologicals Biologicals™

Product Code. 30492166 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30492166 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30492166 Supplier Novus Biologicals Supplier No. NBP337934R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HTF9C Polyclonal antibody specifically detects HTF9C in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTF9C
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias HTF9CHpaII tiny fragments locus 9c protein, MGC102728, TRM2 tRNA methyltransferase 2 homolog A (S. cerevisiae), tRNA (uracil-5-)-methyltransferase homolog A, tRNA methyltransferase 2 homolog A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human HTF9C (NP_073564.3).,, Sequence:, MSENLDNEGPKPMESCGQESSSALSCPTVSVPPAAPAALEEVEKEGAGAATGPGPQPGLYSYIRDDLFTSEIFKLELQNVPRHASFSDVRRFLGRFGLQPHKTKLFGQPPCAFVTFRSAAERDKALRVLHGALWKGRPLSVRLARPKADPMARRRRQEGESEPPVTRVADVVTPLWTVPYAEQLERKQLECEQVLQKLAKEIGSTNRALLPWLLEQRHKHNKACCPLEGVRPSPQQTEYRNKCEFLVGVG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 27037
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.