missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APJ Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8561-SP
This item is not returnable.
View return policy
Description
APJ/Apelin receptor Monoclonal specifically detects APJ/Apelin receptor in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| APJ/Apelin receptor | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AGTRL1, AGTRL1HG11, angiotensin II receptor-like 1, Angiotensin receptor-like 1, apelin receptor, APJ (apelin) receptor, APJ receptor, APJFLJ96609, APJR, APLNR, FLJ90771, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11, HG11 orphan receptor, MGC45246 | |
| Mouse myeloma cell line NS0-derived recombinant human APJ Met1-Asp380 Accession # P35414 | |
| 25 μg | |
| Primary | |
| Detects human APJ in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry | |
| 72133R | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 0.5-25 ug/mL | |
| P35414 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 187 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction