missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APJ Antibody, R&D Systems™

Mouse Monoclonal Antibody
249.00 € - 488.00 €
Specifications
| Antigen | APJ/Apelin receptor |
|---|---|
| Clone | 72133R |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 0.5-25 ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
18734689
![]() |
R&D Systems
MAB8561-SP |
25 μg |
249.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18110051
![]() |
R&D Systems
MAB8561-100 |
100 μg |
488.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
APJ/Apelin receptor Monoclonal specifically detects APJ/Apelin receptor in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifica
| APJ/Apelin receptor | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 0.5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AGTRL1, AGTRL1HG11, angiotensin II receptor-like 1, Angiotensin receptor-like 1, apelin receptor, APJ (apelin) receptor, APJ receptor, APJFLJ96609, APJR, APLNR, FLJ90771, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11, HG11 orphan receptor, MGC45246 | |
| Mouse myeloma cell line NS0-derived recombinant human APJ Met1-Asp380 Accession # P35414 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human APJ in direct ELISAs. |
| 72133R | |
| Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P35414 | |
| 187 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto