missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-1 Adrenergic R/ADRB1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB10119-100
This item is not returnable.
View return policy
Description
beta-1 Adrenergic R/ADRB1 Monoclonal specifically detects beta-1 Adrenergic R/ADRB1 in Human samples. It is validated for Western Blot.
Specifications
| beta-1 Adrenergic R/ADRB1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADRB1, ADRB1R, ADRB1RRHR, adrenergic, beta-1-, receptor, B1AR, beta-1 adrenergic receptor, beta1 AdrenergicR, beta-1 AdrenergicR, Beta-1 adrenoceptor, Beta-1 adrenoreceptor, BETA1AR | |
| Synthetic peptide containing human beta-1 Adrenergic R/ADRB1, Accession No. P08588 | |
| Primary | |
| Detects human beta-1 Adrenergic R/ADRB1 in direct ELISAs. | |
| Human | |
| IgG |
| Western Blot | |
| 2203D | |
| Western Blot 1 μ/mL | |
| P08588 | |
| Rabbit | |
| 100 μg | |
| 153 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction