missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-1 Adrenergic R/ADRB1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
206.00 € - 468.00 €
Specifications
| Antigen | beta-1 Adrenergic R/ADRB1 |
|---|---|
| Clone | 2203D |
| Dilution | Western Blot 1 μ/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18740867
![]() |
R&D Systems
MAB10119-SP |
25 μg |
206.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18723347
![]() |
R&D Systems
MAB10119-100 |
100 μg |
468.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-1 Adrenergic R/ADRB1 Monoclonal specifically detects beta-1 Adrenergic R/ADRB1 in Human samples. It is validated for Western Blot.Specifications
| beta-1 Adrenergic R/ADRB1 | |
| Western Blot 1 μ/mL | |
| Monoclonal | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADRB1, ADRB1R, ADRB1RRHR, adrenergic, beta-1-, receptor, B1AR, beta-1 adrenergic receptor, beta1 AdrenergicR, beta-1 AdrenergicR, Beta-1 adrenoceptor, Beta-1 adrenoreceptor, BETA1AR | |
| Synthetic peptide containing human beta-1 Adrenergic R/ADRB1, Accession No. P08588 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 2203D | |
| Western Blot | |
| Unconjugated | |
| Human | |
| P08588 | |
| 153 | |
| IgG | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human beta-1 Adrenergic R/ADRB1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title