missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR5 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems FAB182F-025
This item is not returnable.
View return policy
Description
CCR5 Monoclonal specifically detects CCR5 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR5 | |
| Monoclonal | |
| Fluorescein | |
| P51681 | |
| CCR5 | |
| NS0 mouse myeloma cell line transfected with human CCR5 Met1-Leu352 Accession # P51681 | |
| 25 Tests | |
| 1234 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 45531 | |
| Flow Cytometry 10 uL/10^6 cells | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human CCR5. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction