missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR5 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
213.00 € - 416.00 €
Specifications
| Antigen | CCR5 |
|---|---|
| Clone | 45531 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722008
![]() |
R&D Systems
FAB182F-025 |
25 Tests |
213.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755436
![]() |
R&D Systems
FAB182F-100 |
100 Tests |
416.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR5 Monoclonal specifically detects CCR5 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR5 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| CCR5 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Detects human CCR5. |
| 45531 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| P51681 | |
| 1234 | |
| NS0 mouse myeloma cell line transfected with human CCR5 Met1-Leu352 Accession # P51681 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title