missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD27 Ligand/TNFSF7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB27381-100
This item is not returnable.
View return policy
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD27 Ligand/TNFSF7/CD70 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
| Mouse myeloma cell line NS0-derived recombinant human CD27 Ligand Gln45-Pro193 Accession # P32970 | |
| 100 μg | |
| Primary | |
| Detects human CD27 Ligand/TNFSF7 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry | |
| 832614 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| P32970 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 970 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction