missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD27 Ligand/TNFSF7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 425.00 €
Specifications
| Antigen | CD27 Ligand/TNFSF7/CD70 |
|---|---|
| Clone | 832614 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18787428
![]() |
R&D Systems
MAB27381-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18735139
![]() |
R&D Systems
MAB27381-100 |
100 μg |
425.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow Cytometry.Specifications
| CD27 Ligand/TNFSF7/CD70 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
| Mouse myeloma cell line NS0-derived recombinant human CD27 Ligand Gln45-Pro193 Accession # P32970 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD27 Ligand/TNFSF7 in direct ELISAs. |
| 832614 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P32970 | |
| 970 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title