missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD28 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3421-100
This item is not returnable.
View return policy
Description
CD28 Monoclonal specifically detects CD28 in Human samples. It is validated for Immunohistochemistry.
Specifications
| CD28 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| Synthetic peptide containing human CD28, Accession No. P10747 | |
| 100 μg | |
| 940.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 987701 | |
| Immunohistochemistry 5-25 μg/mL | |
| P10747 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human CD28 in direct ELISAs. | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction