missing translation for 'onlineSavingsMsg'
Learn More

Human CD28 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

203.00 € - 456.00 €

Specifications

Antigen CD28
Clone 987701
Dilution Immunohistochemistry 5-25 μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18793137
GREENER_CHOICE
View Documents
R&D Systems
MAB3421-SP
25 μg
203.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18740277
GREENER_CHOICE
View Documents
R&D Systems
MAB3421-100
100 μg
456.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD28 Monoclonal specifically detects CD28 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

CD28
Immunohistochemistry 5-25 μg/mL
Monoclonal
Lyophilized
Human
P10747
940.0
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
987701
Immunohistochemistry
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Synthetic peptide containing human CD28, Accession No. P10747
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CD28 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.