missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD28 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB3422-100
This item is not returnable.
View return policy
Description
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry
Specifications
| CD28 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 um filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 um filtered solution in PBS. | |
| CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD28, Asn19-Pro152, Accession # P10747 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry | |
| 2946A | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| P10747 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 940 | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur