missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD28 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
252.00 € - 571.00 €
Specifications
| Antigen | CD28 |
|---|---|
| Clone | 2946A |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794414
![]() |
R&D Systems
MAB3422-SP |
25 μg |
252.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18704544
|
R&D Systems
MAB3422-100 |
100 μg |
571.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow CytometrySpecifications
| CD28 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| P10747 | |
| 940 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
| 2946A | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD28, Asn19-Pro152, Accession # P10747 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title