missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD300a/LMIR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB26401-100
This item is not returnable.
View return policy
Description
CD300a/LMIR1 Monoclonal antibody specifically detects CD300a/LMIR1 in Human samples. It is validated for ELISA
Specifications
| CD300a/LMIR1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11314 | |
| Human | |
| Purified |
| ELISA | |
| 232618 | |
| ELISA | |
| CD300a, CD300a antigenCMRF35H9, CD300a molecule, CLM8, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, IRp60, leukocyte membrane antigen, LMIR1, MAIR-I, Mcipr1, NKRL, Pigr4 | |
| Y3 rat myeloid cell line transfected with human CD300a/LMIR1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction