missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD300c Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB3256-100
This item is not returnable.
View return policy
Description
CD300c2 Monoclonal antibody specifically detects CD300c2 in Human samples. It is validated for nan
Specifications
| CD300c2 | |
| 2799A | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| NS0-derived human CD300c protein, Met29-Arg183, Accession # Q08708 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Monoclonal | |
| Unconjugated | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 140497 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction