missing translation for 'onlineSavingsMsg'
Learn More

Human CD300c Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

191.00 € - 506.00 €

Specifications

Antigen CD300c2
Clone 2799A
Classification Monoclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18723617
GREENER_CHOICE
View Documents
R&D Systems
MAB3256-SP
25 μg
191.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18783137
GREENER_CHOICE
View Documents
R&D Systems
MAB3256-100
100 μg
506.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD300c2 Monoclonal antibody specifically detects CD300c2 in Human samples. It is validated for nan
Specifications

Specifications

CD300c2
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
NS0-derived human CD300c protein, Met29-Arg183, Accession # Q08708
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
2799A
Unconjugated
Rabbit
Human
140497
IgG
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.