missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD45 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB1430R-100UG
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD45 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5788 | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 2D1 | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| PTPRC | |
| Human peripheral blood mononuclear cells | |
| 100 μg | |
| Primary | |
| Detects human CD45. This antibody recognizes all isoforms of human CD45. | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction