missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD45 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
320.00 € - 322.00 €
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 2D1 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18105721
|
R&D Systems
FAB1430R-100 |
100 Tests |
320.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18723658
|
R&D Systems
FAB1430R-100UG |
100 μg |
322.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45 Monoclonal specifically detects CD45 in Human samples. It is validated for Flow Cytometry.Specifications
| CD45 | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| 5788 | |
| Human peripheral blood mononuclear cells | |
| Primary | |
| Reconstitution default not known | |
| Detects human CD45. This antibody recognizes all isoforms of human CD45. |
| 2D1 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Monoclonal | |
| Mouse | |
| Human | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| PTPRC | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title