missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD90/Thy1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems FAB2067A-100
This item is not returnable.
View return policy
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD90/Thy1 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7070 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| Thy-1A1 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P04216 | |
| THY1 | |
| Human CD90/Thy1 Accession # P04216 | |
| 100 Tests | |
| Primary | |
| Detects human CD90/Thy1. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction