missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD90/Thy1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
325.00 € - 507.00 €
Specifications
| Antigen | CD90/Thy1 |
|---|---|
| Clone | Thy-1A1 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18727437
![]() |
R&D Systems
FAB2067A-025 |
25 Tests |
325.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18728358
![]() |
R&D Systems
FAB2067A-100 |
100 Tests |
507.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry.Specifications
| CD90/Thy1 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| THY1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CD90/Thy1. |
| Thy-1A1 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| P04216 | |
| 7070 | |
| Human CD90/Thy1 Accession # P04216 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title