missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CDX2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3665-100
This item is not returnable.
View return policy
Description
CDX2 Monoclonal specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| CDX2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX3, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| E. coli-derived recombinant human CDX2 Asn54-Thr185 Accession # Q99626 | |
| 100 μg | |
| Primary | |
| Detects human CDX2 in direct ELISAs. | |
| Human | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry | |
| 963809 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Q99626 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1045 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction