missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CDX2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
213.00 € - 438.00 €
Specifications
| Antigen | CDX2 |
|---|---|
| Clone | 963809 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18717039
![]() |
R&D Systems
MAB3665-SP |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18712949
![]() |
R&D Systems
MAB3665-100 |
100 μg |
438.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDX2 Monoclonal specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
| CDX2 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q99626 | |
| 1045 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 963809 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX3, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| E. coli-derived recombinant human CDX2 Asn54-Thr185 Accession # Q99626 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CDX2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title