missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CHRM2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6377
This item is not returnable.
View return policy
Description
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| Muscarinic Acetylcholine Receptor M2/CHRM2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| P08172 | |
| CHRM2 | |
| NS0 mouse myeloma cell line transfected with human CHRM2 Accession # P08172 | |
| 100 μg | |
| Primary | |
| Detects human CHRM2 in immunohistochemistry. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 585713 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, mAChR M2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1129 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction