missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CHRM2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 397.00 €
Specifications
| Antigen | Muscarinic Acetylcholine Receptor M2/CHRM2 |
|---|---|
| Clone | 585713 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759698
![]() |
R&D Systems
MAB6377-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179870
|
R&D Systems
MAB6377 |
100 μg |
397.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| Muscarinic Acetylcholine Receptor M2/CHRM2 | |
| LYOPH | |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08172 | |
| 1129 | |
| NS0 mouse myeloma cell line transfected with human CHRM2 Accession # P08172 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human CHRM2 in immunohistochemistry. |
| 585713 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, mAChR M2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor | |
| CHRM2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title