missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Common gamma Chain/IL-2 R gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2841
This item is not returnable.
View return policy
Description
Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Common gamma Chain/IL-2 R gamma | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 R gamma, IL-2 receptor subunit gamma, IL2R gamma, IL-2R subunit gamma, IL2RG, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3561 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 31134 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P31785 | |
| IL2RG | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Common gamma Chain/IL-2 R gamma Leu23-Asn254 Accession # P31785 | |
| 500 μg | |
| Primary | |
| Detects human Common gamma Chain/IL-2 R gamma in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction