missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Common gamma Chain/IL-2 R gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
188.00 € - 500.00 €
Specifications
| Antigen | Common gamma Chain/IL-2 R gamma |
|---|---|
| Clone | 31134 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18775138
![]() |
R&D Systems
MAB2841-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18190711
![]() |
R&D Systems
MAB2841 |
500 μg |
500.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| Common gamma Chain/IL-2 R gamma | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 R gamma, IL-2 receptor subunit gamma, IL2R gamma, IL-2R subunit gamma, IL2RG, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
| IL2RG | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 31134 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P31785 | |
| 3561 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Common gamma Chain/IL-2 R gamma Leu23-Asn254 Accession # P31785 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Common gamma Chain/IL-2 R gamma in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title