Learn More
Human Corin Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB2209-500
 View more versions of this product
Product Code. 18740658
  • 2160.00 € / 500µg
Description
Corin Monoclonal specifically detects Corin in Human samples. It is validated for ELISA, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Corin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ATC2, atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
10699
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Western Blot, Immunocytochemistry
231443
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Q9Y5Q5
CORIN
Mouse myeloma cell line NS0-derived recombinant human Corin Ser67-Asn1042 Accession # Q9Y5Q5
500 μg
Primary
Detects human Corin in direct ELISAs and Western blots.
Human
Supernatant