missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Corin Antibody, R&D Systems™

Rat Monoclonal Antibody
221.00 € - 2160.00 €
Specifications
| Antigen | Corin |
|---|---|
| Clone | 231443 |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752509
![]() |
R&D Systems
MAB2209-SP |
25 μg |
221.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773808
![]() |
R&D Systems
MAB2209-100 |
100 μg |
519.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18740658
![]() |
R&D Systems
MAB2209-500 |
500 μg |
2160.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Corin Monoclonal specifically detects Corin in Human samples. It is validated for ELISA, Immunocytochemistry.Specifications
| Corin | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Rat | |
| Human | |
| Q9Y5Q5 | |
| 10699 | |
| Mouse myeloma cell line NS0-derived recombinant human Corin Ser67-Asn1042 Accession # Q9Y5Q5 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Corin in direct ELISAs and Western blots. |
| 231443 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ATC2, atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10 | |
| CORIN | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title