Learn More
Human Cytokeratin 17 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11571-100
 View more versions of this product
Product Code. 30245516
  • 536.00 € / 100µg
Description
Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human samples. It is validated for Immunocytochemistry,Western Blot,Simple Western,Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 17
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin 17, type I, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1
Cytokeratin 17 containing synthetic peptide, Accession # Q04695
100 μg
Primary
Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution.
IgG2b
Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry
1080235
Western Blot 2 μg/mL, Simple Western 100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL
Q04695
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3872
Human
Purified