Learn More
Human DAP12 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB52401-SP
 View more versions of this product
Product Code. 18771997
  • 189.00 € / 25µg
Description
DAP12 Monoclonal antibody specifically detects DAP12 in Human samples. It is validated for nan
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DAP12
406209
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7305
Human
Purified
Monoclonal
Unconjugated
DAP12PLOSL, DNAX-activation protein 12, KARAPPLO-SL, KAR-associated protein, killer activating receptor associated protein, Killer-activating receptor-associated protein, TYRO protein tyrosine kinase binding protein, TYRO protein tyrosine kinase-binding protein
Human Dap12 synthetic peptide, Accession # O43914
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a