missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DAP12 Antibody, R&D Systems™

Mouse Monoclonal Antibody
189.00 € - 497.00 €
Specifications
| Antigen | DAP12 |
|---|---|
| Clone | 406209 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18771997
![]() |
R&D Systems
MAB52401-SP |
25 μg |
189.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702687
![]() |
R&D Systems
MAB52401-100 |
100 μg |
497.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DAP12 Monoclonal antibody specifically detects DAP12 in Human samples. It is validated for nanSpecifications
| DAP12 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 7305 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 406209 | |
| Unconjugated | |
| Mouse | |
| Human | |
| DAP12PLOSL, DNAX-activation protein 12, KARAPPLO-SL, KAR-associated protein, killer activating receptor associated protein, Killer-activating receptor-associated protein, TYRO protein tyrosine kinase binding protein, TYRO protein tyrosine kinase-binding protein | |
| Human Dap12 synthetic peptide, Accession # O43914 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title