missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DC-SIGN+DC-SIGNR Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
209.00 € - 572.00 €
Specifications
| Antigen | DC-SIGN+DC-SIGNR |
|---|---|
| Clone | DC28 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade 4.0 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792737
![]() |
R&D Systems
MAB16211-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18170461
![]() |
R&D Systems
MAB16211 |
500 μg |
572.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DC-SIGN+DC-SIGNR Monoclonal specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready.Specifications
| DC-SIGN+DC-SIGNR | |
| LYOPH | |
| Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| 30835 | |
| E. coli-derived recombinant human DC-SIGN Extracellular domain | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human DC-SIGN in direct ELISAs and Western blots. Was reported to cross-react with human DC-SIGNR as well as DC-SIGN from Pigtailed Macaque and Rhesus Macaque (10). |
| DC28 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade 4.0 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD209 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title