missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EDA-A2/Ectodysplasin A2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF922-SP
This item is not returnable.
View return policy
Description
EDA-A2/Ectodysplasin A2 Polyclonal specifically detects EDA-A2/Ectodysplasin A2 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.
Specifications
| EDA-A2/Ectodysplasin A2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EDA | |
| Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2 Ala179-Ser389 Accession # NP_001005609 | |
| 25 μg | |
| Primary | |
| Detects human EDA-A2/Ectodysplasin A2 in direct ELISAs and Western blots. In Western blots, less than 10% cross-reactivity with recombinant human (rh) APRIL, rhBAFF, rhLIGHT, rhOX40 Ligand, and rhTRAIL is observed and less than 2% cross-reactivity with recombinant mouse EDA, rhTNF-alpha , rhVEGI, rhGITR Ligand, and rhFasLigand is observed. | |
| Human | |
| IgG |
| Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Blockade of Receptor-ligand Interaction | |
| NP_001005609 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1896 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction