missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EDA-A2/Ectodysplasin A2 Antibody, R&D Systems™

Goat Polyclonal Antibody
188.00 € - 446.00 €
Specifications
| Antigen | EDA-A2/Ectodysplasin A2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Blockade of Receptor-ligand Interaction |
| Applications | Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785097
![]() |
R&D Systems
AF922-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18052582
![]() |
R&D Systems
AF922 |
100 μg |
446.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EDA-A2/Ectodysplasin A2 Polyclonal specifically detects EDA-A2/Ectodysplasin A2 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
| EDA-A2/Ectodysplasin A2 | |
| Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Human | |
| 1896 | |
| Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2 Ala179-Ser389 Accession # NP_001005609 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human EDA-A2/Ectodysplasin A2 in direct ELISAs and Western blots. In Western blots, less than 10% cross-reactivity with recombinant human (rh) APRIL, rhBAFF, rhLIGHT, rhOX40 Ligand, and rhTRAIL is observed and less than 2% cross-reactivity with recombinant mouse EDA, rhTNF-alpha , rhVEGI, rhGITR Ligand, and rhFasLigand is observed. |
| LYOPH | |
| Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| NP_001005609 | |
| EDA | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title