missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RIIB/C (CD32b/c) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB18751-100
This item is not returnable.
View return policy
Description
Fc gamma RIIB/C (CD32b/c) Monoclonal specifically detects Fc gamma RIIB/C (CD32b/c) in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Fc gamma RIIB/C (CD32b/c) | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P31994 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2213 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 190710 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FCGR2B | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIB/C Ala46-Pro217 Accession # P31994 | |
| 100 μg | |
| Primary | |
| Detects human Fc gamma RIIB/C in ELISAs. In a sandwich ELISA shows approximately 40% cross-reactivity with recombinant human (rh) Fc gamma RIIA and no cross-reactivity with rhFc gamma RI, rmFc gamma RI, rmFc gamma RIIB, rhFc gamma RIIIB, rmFc gamma RIII, or rmFcrl3/CD16-2. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction