missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RIIB/C (CD32b/c) Antibody, R&D Systems™

Mouse Monoclonal Antibody
211.00 € - 2050.00 €
Specifications
| Antigen | Fc gamma RIIB/C (CD32b/c) |
|---|---|
| Clone | 190710 |
| Concentration | LYOPH |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762559
![]() |
R&D Systems
MAB18751-SP |
25 μg |
211.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18770187
![]() |
R&D Systems
MAB18751-100 |
100 μg |
528.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18787569
![]() |
R&D Systems
MAB18751-500 |
500 μg |
2050.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIIB/C (CD32b/c) Monoclonal specifically detects Fc gamma RIIB/C (CD32b/c) in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Fc gamma RIIB/C (CD32b/c) | |
| LYOPH | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2213 | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIB/C Ala46-Pro217 Accession # P31994 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Fc gamma RIIB/C in ELISAs. In a sandwich ELISA shows approximately 40% cross-reactivity with recombinant human (rh) Fc gamma RIIA and no cross-reactivity with rhFc gamma RI, rmFc gamma RI, rmFc gamma RIIB, rhFc gamma RIIIB, rmFc gamma RIII, or rmFcrl3/CD16-2. |
| 190710 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P31994 | |
| FCGR2B | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title