missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RIIIA/B (CD16) Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF1597-SP
This item is not returnable.
View return policy
Description
Fc gamma RIII (CD16) Polyclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| Fc gamma RIII (CD16) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2214 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction | |
| O75015 | |
| FCGR3B | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIIB (R&D Systems, Catalog # 1597-FC) Thr20-Gln208 Accession # O75015 | |
| 25 μg | |
| Primary | |
| Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human Fc gamma RIIA is observed and less than 2% cross-reactivity with recombinant mouse CD16 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction