missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RIIIA/B (CD16) Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
226.00 € - 605.00 €
Specifications
| Antigen | Fc gamma RIII (CD16) |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705967
![]() |
R&D Systems
AF1597-SP |
25 μg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18090562
|
R&D Systems
AF1597 |
100 μg |
605.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIII (CD16) Polyclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
| Fc gamma RIII (CD16) | |
| Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Human | |
| CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20 | |
| FCGR3B | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| O75015 | |
| 2214 | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIIB (R&D Systems, Catalog # 1597-FC) Thr20-Gln208 Accession # O75015 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human Fc gamma RIIA is observed and less than 2% cross-reactivity with recombinant mouse CD16 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title