missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibrinogen Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9404-SP
This item is not returnable.
View return policy
Description
Fibrinogen Monoclonal antibody specifically detects Fibrinogen in Human samples. It is validated for Immunocytochemistry.
Specifications
| Fibrinogen | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Factor I, Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
| Human serum-derived Fibrinogen, Accession # P02671 | |
| 25 μg | |
| Primary | |
| Detects human Fibrinogen in direct ELISAs. In Western blots, does not detect Fibrinogen A. | |
| Human | |
| IgG1 |
| Immunocytochemistry | |
| 949332 | |
| Immunocytochemistry 8-25 ug/mL | |
| P02671 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2243 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction