missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibrinogen Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 417.00 €
Specifications
| Antigen | Fibrinogen |
|---|---|
| Clone | 949332 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772989
![]() |
R&D Systems
MAB9404-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732618
![]() |
R&D Systems
MAB9404-100 |
100 μg |
417.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibrinopeptide B Monoclonal specifically detects Fibrinopeptide B in Human samples. It is validated for Immunocytochemistry.Specifications
| Fibrinogen | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P02671 | |
| 2243 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 949332 | |
| Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Factor I, Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
| Human serum-derived Fibrinogen, Accession # P02671 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Fibrinogen in direct ELISAs. In Western blots, does not detect Fibrinogen A. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title