missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gamma-Synuclein Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5745R-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
gamma-Synuclein Monoclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Especificaciones
| gamma-Synuclein | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BCSG1, BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, PRSN, SNCG, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1), Synuclein-gamma | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6623 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 514304R | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 1-25 ug/mL | |
| Q6FHG5 | |
| SNCG | |
| E. coli-derived recombinant human gamma-Synuclein Asn47-Glu120 Accession # Q6FHG5 | |
| 25 μg | |
| Primary | |
| Detects human gamma-Synuclein in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido