missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gamma-Synuclein Antibody, R&D Systems™

Mouse Monoclonal Antibody
226.00 € - 2150.00 €
Specifications
| Antigen | gamma-Synuclein |
|---|---|
| Clone | 514304R |
| Dilution | Western Blot 0.5 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 1-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776068
![]() |
R&D Systems
MAB5745R-SP |
25 μg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762798
![]() |
R&D Systems
MAB5745R-100 |
100 μg |
518.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18136091
|
R&D Systems
MAB5745R-01M |
1 mg |
2035.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141891
|
R&D Systems
MAB5745R-500 |
500 ug |
2150.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gamma-Synuclein Monoclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| gamma-Synuclein | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q6FHG5 | |
| 6623 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Detects human gamma-Synuclein in direct ELISAs and Western blots. |
| 514304R | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| BCSG1, BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, PRSN, SNCG, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1), Synuclein-gamma | |
| SNCG | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title