Learn More
Human GHRHR Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB10735-SP
 View more versions of this product
Product Code. 18711237
  • 201.00 € / 25µg
Description
GHRHR Monoclonal antibody specifically detects GHRHR in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GHRHR
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
GHRFR, GHRH receptor, GRF receptor, GRFR, growth hormone releasing hormone receptor, Growth hormone-releasing factor receptor, growth hormone-releasing hormone receptor, IGHD1B, Little
Human GHRHR synthetic peptide, Accession # Q02643
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2a
Immunohistochemistry
1037409
Immunohistochemistry 5-25 ug/mL
Q02643
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2692
Human
Purified