missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GHRHR Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 € - 428.00 €
Specifications
| Antigen | GHRHR |
|---|---|
| Clone | 1037409 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711237
![]() |
R&D Systems
MAB10735-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790947
![]() |
R&D Systems
MAB10735-100 |
100 μg |
428.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GHRHR Monoclonal antibody specifically detects GHRHR in Human samples. It is validated for ImmunohistochemistrySpecifications
| GHRHR | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| GHRFR, GHRH receptor, GRF receptor, GRFR, growth hormone releasing hormone receptor, Growth hormone-releasing factor receptor, growth hormone-releasing hormone receptor, IGHD1B, Little | |
| Human GHRHR synthetic peptide, Accession # Q02643 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1037409 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q02643 | |
| 2692 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title