missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GLI-3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB3690-100
This item is not returnable.
View return policy
Description
GLI-3 Monoclonal antibody specifically detects GLI-3 in Human samples. It is validated for Immunocytochemistry
Specifications
| GLI-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ACLS, ACLSPAPAPHS, ADD, Bph, GCPS, GLI family zinc finger 3, GLI3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPB, PAPBDNA-binding protein, Pdn, PHS, PPDIV, Xt, zinc finger protein GLI3 | |
| E. coli-derived recombinant human GLI-3, Met1-Glu479, Accession # P10071 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Immunocytochemistry | |
| 2352B | |
| Immunocytochemistry 3-25 ug/mL | |
| P10071 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2737 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction