missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GLI-3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
252.00 € - 574.00 €
Specifications
| Antigen | GLI-3 |
|---|---|
| Clone | 2352B |
| Dilution | Immunocytochemistry 3-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710907
![]() |
R&D Systems
MAB3690-SP |
25 μg |
252.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732877
![]() |
R&D Systems
MAB3690-100 |
100 μg |
574.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLI-3 Monoclonal antibody specifically detects GLI-3 in Human samples. It is validated for ImmunocytochemistrySpecifications
| GLI-3 | |
| Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ACLS, ACLSPAPAPHS, ADD, Bph, GCPS, GLI family zinc finger 3, GLI3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPB, PAPBDNA-binding protein, Pdn, PHS, PPDIV, Xt, zinc finger protein GLI3 | |
| E. coli-derived recombinant human GLI-3, Met1-Glu479, Accession # P10071 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2352B | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P10071 | |
| 2737 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title