missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Haptoglobin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8465-100
This item is not returnable.
View return policy
Description
Haptoglobin Monoclonal specifically detects Haptoglobin in Human samples. It is validated for ELISA.
Specifications
| Haptoglobin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP, HP-1, hp2-alpha, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
| Mouse myeloma cell line NS0-derived recombinant human Haptoglobin, Met1-Gln347, Accession No. P00738 | |
| 100 μg | |
| 3240.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 448018 | |
| ELISA | |
| P00738 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Haptoglobin in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction