missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Haptoglobin Antibody, R&D Systems™

Mouse Monoclonal Antibody
194.00 € - 487.00 €
Specifications
| Antigen | Haptoglobin |
|---|---|
| Clone | 448018 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721157
![]() |
R&D Systems
MAB8465-SP |
25 μg |
194.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752197
![]() |
R&D Systems
MAB8465-100 |
100 μg |
487.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Haptoglobin Monoclonal specifically detects Haptoglobin in Human samples. It is validated for ELISA.Specifications
| Haptoglobin | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P00738 | |
| 3240.0 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 448018 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP, HP-1, hp2-alpha, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
| Mouse myeloma cell line NS0-derived recombinant human Haptoglobin, Met1-Gln347, Accession No. P00738 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Haptoglobin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title